Assay Details
Target Gene Details
Entrez Gene ID: | 16549 |
Gene Name: | KH-type splicing regulatory protein |
Gene Aliases: |
6330409F21Rik, Fbp2, Fubp2, Ksrp |
Location: |
Chr.17:57021049-57031507 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Khsrp | NM_010613.3 | NP_034743.3 | ||
| AK077415.1 | ||||
| AK156541.1 | BAE33750.1 | |||
| BB645370.1 | ||||
| BY138109.1 |
More Information
Set Membership: |
|

Genomic Map