Assay Details
Target Gene Details
Entrez Gene ID: | 26407 |
Gene Name: | mitogen-activated protein kinase kinase kinase 4 |
Gene Aliases: |
D17Rp17, D17Rp17e, MAPKKK4, MEKK 4, MTK1, Mek4b, Mekk4, RP17, Rp17a, Tas, mKIAA0213 |
Location: |
Chr.17:12227621-12318806 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k4 | NM_011948.2 | 11 | 2948 | NP_036078.2 |
| AK122219.1 | 10 | 2631 | BAC65501.1 | |
| AK133605.1 | 11 | 3085 | BAE21746.1 | |
| BC058719.1 | 11 | 2948 | AAH58719.1 | |
| U66240.1 | 9 | 1831 | AAC08286.1 | |
| U85607.1 | 11 | 3036 | AAC53126.1 | |
| U85608.1 | 11 | 3036 | AAC53127.1 |
More Information
Set Membership: |
|

Genomic Map