Assay Details
Target Gene Details
Entrez Gene ID: | 67938 |
Gene Name: | myosin, light chain 12B, regulatory |
Gene Aliases: |
1500001M02Rik, C77744, Mylc2b, RLC-B |
Location: |
Chr.17:70973963-70990516 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myl12b | NM_023402.2 | NP_075891.1 | ||
| AK002885.1 | BAB22432.1 | |||
| AK005133.1 | BAB23832.1 | |||
| AK146115.1 | BAE26912.1 | |||
| AK160123.1 | BAE35646.1 | |||
| AK168316.1 | BAE40255.1 | |||
| BC028878.1 | AAH28878.1 | |||
| BC131925.1 | ||||
| BC131927.1 |
More Information
Set Membership: |
|

Genomic Map