Assay Details
Target Gene Details
Entrez Gene ID: | 71893 |
Gene Name: | NADPH oxidase organizer 1 |
Gene Aliases: |
2310034C04Rik, P41NOX, P41NOXA, P41NOXB, P41NOXC, Snx28, hslt |
Location: |
Chr.17:24696234-24700530 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Noxo1 | NM_027988.4 | NP_082264.2 | ||
| AF399754.1 | AAK94017.1 | |||
| AF539797.1 | AAN75142.1 | |||
| AK009605.1 | 6 | 795 | BAB26387.1 | |
| AK088226.1 | BAC40222.1 | |||
| AK157993.1 | BAE34305.1 | |||
| AK184735.1 | ||||
| BC019525.1 | AAH19525.1 | |||
| BC119527.1 | ||||
| BC119528.1 | ||||
| BC127235.1 | ||||
| BC127236.1 |

Genomic Map