Assay Details
Target Gene Details
Entrez Gene ID: | 68531 |
Gene Name: | RIKEN cDNA 1110020A21 gene |
Gene Aliases: |
2600015P10Rik |
Location: |
Chr.17:84917182-84957710 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110020A21Rik | NR_027929.1 | |||
| NR_027930.1 | ||||
| AK003841.1 | ||||
| AK011222.1 | ||||
| BI737205.1 |
Target Gene Details
Entrez Gene ID: | 19043 |
Gene Name: | protein phosphatase 1B, magnesium dependent, beta isoform |
Gene Aliases: |
PP2CB |
Location: |
Chr.17:84956741-85023992 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1b | NM_001159497.1 | 1 | 111 | NP_001152969.1 |
| AK076923.1 | 1 | 114 | ||
| BB615769.1 | 1 | 112 |
More Information
Set Membership: |
|

Genomic Map