Assay Details
Target Gene Details
Entrez Gene ID: | 19043 |
Gene Name: | protein phosphatase 1B, magnesium dependent, beta isoform |
Gene Aliases: |
PP2CB |
Location: |
Chr.17:84956741-85023992 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1b | NM_001159496.1 | NP_001152968.1 | ||
| NM_001159497.1 | NP_001152969.1 | |||
| NM_001159498.1 | NP_001152970.1 | |||
| NM_011151.2 | NP_035281.1 | |||
| AJ271833.1 | CAC28024.1 | |||
| AJ271836.1 | CAC28025.1 | |||
| AK010224.1 | BAE43218.1 | |||
| AK050170.1 | ||||
| AK076923.1 | ||||
| AK077136.1 | ||||
| AK136077.1 | BAE22808.1 | |||
| AK160653.1 | ||||
| AK163283.1 | ||||
| AK167176.1 | BAE39311.1 | |||
| BC018556.1 | AAH18556.1 | |||
| BC090963.1 | AAH90963.1 | |||
| BQ176513.1 | ||||
| D17411.1 | BAA04233.1 | |||
| D17412.1 | BAA04234.1 | |||
| D45859.1 | BAA08293.1 | |||
| D45860.1 | BAA08294.1 | |||
| D45861.1 | BAA08295.1 | |||
| U09218.1 | AAB60442.1 |

Genomic Map