Assay Details
Target Gene Details
Entrez Gene ID: | 22153 |
Gene Name: | tubulin, beta 4A class IVA |
Gene Aliases: |
AI325297, M(beta)4, Tubb, Tubb4 |
Location: |
Chr.17:57080066-57087782 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb4a | NM_009451.3 | NP_033477.2 | ||
| AK013717.1 | BAB28967.1 | |||
| BC049112.1 | AAH49112.1 | |||
| BC054831.1 | AAH54831.1 |
More Information
Set Membership: |
|

Genomic Map