Assay Details
Target Gene Details
Entrez Gene ID: | 67864 |
Gene Name: | Yip1 domain family, member 4 |
Gene Aliases: |
2310034L04Rik |
Location: |
Chr.17:74489493-74500277 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yipf4 | NM_026417.4 | 3 | 544 | NP_080693.2 |
| AK009618.1 | 3 | 530 | ||
| AK017721.1 | 3 | 522 | ||
| AK017722.1 | 3 | 522 | ||
| AK083301.1 | 3 | 524 | BAC38852.1 | |
| AK159227.1 | 3 | 505 | BAE34913.1 | |
| AK166347.1 | 3 | 532 | BAE38721.1 | |
| BC003936.1 | 4 | 719 | AAH03936.1 | |
| BY709789.1 | 3 | 544 |

Genomic Map