Assay Details
Target Gene Details
Entrez Gene ID: | 22154 |
Gene Name: | tubulin, beta 5 class I |
Gene Aliases: |
AA408537, AI596182, B130022C14Rik, M(beta)5 |
Location: |
Chr.17:35833920-35838301 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb5 | NM_011655.5 | NP_035785.1 | ||
| AK010960.1 | BAB27292.1 | |||
| AK011263.1 | BAB27504.1 | |||
| AK011765.1 | ||||
| AK012350.1 | ||||
| AK017806.1 | ||||
| AK051164.1 | BAC34541.1 | |||
| AK051393.1 | BAC34623.1 | |||
| AK076196.1 | ||||
| AK083327.1 | BAC38866.1 | |||
| AK146567.1 | BAE27265.1 | |||
| AK148978.1 | BAE28709.1 | |||
| AK151215.1 | BAE30210.1 | |||
| AK151670.1 | BAE30596.1 | |||
| AK160225.1 | BAE35700.1 | |||
| AK161575.1 | BAE36471.1 | |||
| AK165249.1 | BAE38103.1 | |||
| AK167691.1 | BAE39738.1 | |||
| AK167779.1 | BAE39811.1 | |||
| AK167808.1 | BAE39835.1 | |||
| AK167926.1 | BAE39931.1 | |||
| AK168271.1 | BAE40217.1 | |||
| AK169123.1 | BAE40903.1 | |||
| AK169210.1 | BAE40982.1 | |||
| AK169295.1 | BAE41051.1 | |||
| AK169606.1 | ||||
| AK188622.1 | ||||
| AK209234.1 | ||||
| BC003825.1 | AAH03825.1 | |||
| X04663.1 | CAA28369.1 |
More Information
Set Membership: |
|

Genomic Map