Assay Details
Target Gene Details
Entrez Gene ID: | 20383 |
Gene Name: | serine/arginine-rich splicing factor 3 |
Gene Aliases: |
AL024116, Sfrs3, X16 |
Location: |
Chr.17:29032660-29043372 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf3 | NM_013663.5 | NP_038691.1 | ||
| NR_036613.1 | ||||
| AK009895.1 | BAB26569.1 | |||
| AK011657.1 | BAB27762.1 | |||
| AK078891.1 | BAC37445.1 | |||
| AK085863.1 | ||||
| AK088122.1 | ||||
| AK134177.1 | BAE22040.1 | |||
| AK135298.1 | BAE22483.1 | |||
| AK141440.1 | ||||
| AK145089.1 | BAE26230.1 | |||
| AK145600.1 | ||||
| AK145715.1 | BAE26606.1 | |||
| AK146010.1 | BAE26830.1 | |||
| AK146253.1 | BAE27015.1 | |||
| AK146610.1 | ||||
| AK150912.1 | BAE29950.1 | |||
| AK152411.1 | BAE31197.1 | |||
| AK152704.1 | ||||
| AK152779.1 | BAE31490.1 | |||
| AK153073.1 | BAE31698.1 | |||
| AK153209.1 | BAE31808.1 | |||
| AK154046.1 | ||||
| AK178254.1 | ||||
| AK182348.1 | ||||
| BC068111.1 | AAH68111.1 | |||
| BC071196.1 | AAH71196.1 | |||
| BC083316.1 | AAH83316.1 | |||
| BY249648.1 | ||||
| X53824.1 | CAA37821.1 |
More Information
Set Membership: |
|

Genomic Map