Assay Details
Target Gene Details
Entrez Gene ID: | 225027 |
Gene Name: | serine/arginine-rich splicing factor 7 |
Gene Aliases: |
35kDa, 9430065L19Rik, 9G8, NX-96, Sfrs7 |
Location: |
Chr.17:80200080-80207318 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf7 | NM_001195485.1 | NP_001182414.1 | ||
| NM_001195486.1 | NP_001182415.1 | |||
| NM_001195487.1 | NP_001182416.1 | |||
| NM_146083.2 | NP_666195.1 | |||
| NR_036615.1 | ||||
| AK034952.1 | ||||
| AK082848.1 | BAC38650.1 | |||
| AK137795.1 | BAE23499.1 | |||
| AK161652.1 | ||||
| AK168355.1 | BAE40291.1 | |||
| BC014857.1 | AAH14857.1 | |||
| BC025529.1 | AAH25529.1 | |||
| BC027391.1 | AAH27391.1 | |||
| BY740981.1 |
More Information
Set Membership: |
|

Genomic Map