Assay Details
Target Gene Details
Entrez Gene ID: | 386612 |
Gene Name: | THO complex 6 homolog (Drosophila) |
Gene Aliases: |
F830014G06Rik, Wdr58 |
Location: |
Chr.17:23668618-23673903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thoc6 | NM_001008425.1 | NP_001008425.1 | ||
| AK089746.1 | ||||
| BC085131.1 | AAH85131.1 | |||
| BC089551.1 | AAH89551.1 |

Genomic Map