Assay Details
Target Gene Details
Entrez Gene ID: | 21769 |
Gene Name: | zinc finger, AN1-type domain 3 |
Gene Aliases: |
AW539211, TEG-27, Tex27 |
Location: |
Chr.17:30005039-30210020 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfand3 | NM_148926.2 | 3 | 353 | NP_683728.1 |
| AK082915.1 | 2 | 349 | ||
| AK160675.1 | 3 | 212 | ||
| AK181546.1 | 2 | 143 | ||
| AK183489.1 | 2 | 143 | ||
| AK183610.1 | 2 | 143 | ||
| AK184062.1 | 2 | 143 | ||
| AK193183.1 | 1 | 143 | ||
| BC027161.1 | 1 | 85 | AAH27161.1 | |
| BC034396.1 | 3 | 273 | AAH34396.1 | |
| BC083124.1 | 3 | 353 | AAH83124.1 | |
| BC100559.1 | 3 | 204 | AAI00560.1 | |
| EF437371.1 | 1 | 51 | ||
| X80437.1 | 3 | 183 |

Genomic Map