Assay Details
Target Gene Details
Entrez Gene ID: | 224585 |
Gene Name: | zinc finger protein 160 |
Gene Aliases: |
6720480D16Rik, F11, HKr18, HZF5, KR18, mKIAA1611, mszf47 |
Location: |
Chr.17:21008941-21028857 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp160 | NM_145483.2 | NP_663458.2 | ||
| AK143313.1 | BAE25340.1 | |||
| AK173216.1 | BAD32494.1 | |||
| BC015291.1 | AAH15291.1 |
More Information
Set Membership: |
|

Genomic Map