Assay Details
Target Gene Details
Entrez Gene ID: | 225523 |
Gene Name: | centrosomal protein 120 |
Gene Aliases: |
A230075C01, AU016693, Ccdc100 |
Location: |
Chr.18:53681723-53745603 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep120 | NM_178686.3 | NP_848801.2 | ||
| AK038916.1 | BAC30167.1 | |||
| AK044808.1 | BAC32100.1 | |||
| AK047594.1 | BAC33092.1 | |||
| AK079319.1 | BAC37606.1 | |||
| AK089236.1 | ||||
| AK089242.1 | ||||
| BC046493.1 | AAH46493.1 | |||
| BC053439.1 | AAH53439.1 |

Genomic Map