Assay Details
Target Gene Details
Entrez Gene ID: | 12683 |
Gene Name: | cell death-inducing DNA fragmentation factor, alpha subunit-like effector A |
Gene Aliases: |
AW212747 |
Location: |
Chr.18:67343564-67367794 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cidea | NM_007702.2 | 3 | 530 | NP_031728.2 |
| AF041376.1 | 3 | 504 | AAC34985.1 | |
| BB614324.1 | 3 | 305 | ||
| BC096649.1 | 3 | 366 | AAH96649.1 | |
| BQ554477.1 |

Genomic Map