Assay Details
Target Gene Details
Entrez Gene ID: | 12683 |
Gene Name: | cell death-inducing DNA fragmentation factor, alpha subunit-like effector A |
Gene Aliases: |
AW212747 |
Location: |
Chr.18:67343564-67367794 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cidea | NM_007702.2 | 4 | 659 | NP_031728.2 |
| AF041376.1 | 4 | 633 | AAC34985.1 | |
| BB614324.1 | 4 | 434 | ||
| BC096649.1 | 4 | 495 | AAH96649.1 | |
| BQ554476.1 | 2 | 506 |

Genomic Map