Assay Details
Target Gene Details
Entrez Gene ID: | 106628 |
Gene Name: | thyroid hormone receptor interactor 10 |
Gene Aliases: |
AI646975, Cip4 |
Location: |
Chr.17:57233207-57263697 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip10 | NM_001242389.1 | 4 | 407 | NP_001229318.1 |
| NM_001242390.1 | 4 | 407 | NP_001229319.1 | |
| NM_001242391.1 | 4 | 407 | NP_001229320.1 | |
| NM_134125.4 | 4 | 407 | NP_598886.1 | |
| AF502565.1 | 4 | 310 | AAN38709.1 | |
| AK088909.1 | 4 | 357 | BAC40648.1 | |
| AK149902.1 | 4 | 379 | BAE29155.1 | |
| AY081142.1 | 4 | 358 | AAL89589.1 | |
| BC003249.1 | 4 | 409 | AAH03249.1 |

Genomic Map