Assay Details
Target Gene Details
Entrez Gene ID: | 22339 |
Gene Name: | vascular endothelial growth factor A |
Gene Aliases: |
Vegf, Vpf |
Location: |
Chr.17:46016993-46032377 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vegfa | NM_001025250.3 | 3 | 1288 | NP_001020421.2 |
| NM_001025257.3 | 3 | 1288 | NP_001020428.2 | |
| NM_001287056.1 | 3 | 1288 | NP_001273985.1 | |
| NM_001287057.1 | 3 | 1288 | NP_001273986.1 | |
| NM_001287058.1 | 3 | 1288 | NP_001273987.1 | |
| NM_009505.4 | 3 | 1288 | NP_033531.3 | |
| AY120866.1 | 1 | 166 | AAM55477.1 | |
| AY263146.1 | 3 | 267 | AAP86647.1 | |
| AY707864.1 | 4 | 485 | AAU11325.1 | |
| AY750956.1 | 4 | 446 | AAU95484.1 | |
| AY750957.1 | 4 | 409 | AAU95485.1 | |
| AY756068.1 | 3 | 224 | AAV32504.1 | |
| BC061468.1 | 3 | 661 | AAH61468.1 | |
| BQ832724.1 | 2 | 189 | ||
| CA321456.1 | 3 | 661 | ||
| EF028705.1 | 4 | 481 | ||
| M95200.1 | 3 | 353 | AAA40547.1 | |
| S37052.1 | 3 | 279 | AAB22252.1 | |
| S38083.1 | 3 | 279 | AAB22253.1 | |
| S38100.1 | 3 | 279 | AAB22254.1 | |
| U50279.1 | 3 | 267 | AAC05442.1 |
More Information
Set Membership: |
|

Genomic Map