Assay Details
Target Gene Details
Entrez Gene ID: | 67526 |
Gene Name: | autophagy related 12 |
Gene Aliases: |
4931423H11Rik, A330058M13Rik, Apg12l, Atg12l |
Location: |
Chr.18:46732417-46741839 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg12 | NM_026217.3 | NP_080493.2 | ||
| AB066216.1 | BAB62092.1 | |||
| AK005405.1 | BAB24005.1 | |||
| AK008698.1 | BAB25839.1 | |||
| AK016474.2 | BAB30256.1 | |||
| AK039278.1 | 1 | 166 | ||
| AK039546.1 | 1 | 189 | ||
| AK167027.1 | BAE39200.1 | |||
| AK196572.1 | ||||
| BB853539.1 | ||||
| BC070470.1 | AAH70470.1 |

Genomic Map