Assay Details
Target Gene Details
Entrez Gene ID: | 71132 |
Gene Name: | calcium-binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2) |
Gene Aliases: |
1700016C01Rik, 4933421A18Rik, CBP86, FSP-2 |
Location: |
Chr.18:12741307-12755151 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cabyr | NM_001042418.1 | 5 | 1506 | NP_001035883.1 |
| NM_001042419.1 | 5 | 682 | NP_001035884.1 | |
| NM_001042420.1 | NP_001035885.1 | |||
| NM_027687.2 | 5 | 1989 | NP_081963.1 | |
| NM_181731.3 | 5 | 1165 | NP_859420.2 | |
| AF359382.1 | 5 | 1989 | AAK49987.1 | |
| AF359383.1 | 5 | 1165 | AAK49988.1 | |
| AF359384.1 | 5 | 682 | AAK49989.1 | |
| AF359385.1 | 5 | 1506 | AAK49990.1 | |
| AK016856.1 | BAB30467.1 | |||
| AK075751.1 | 5 | 1171 | ||
| AK132770.1 | 5 | 1998 | ||
| BB017109.1 | 1 | 80 | ||
| BC107271.1 | 5 | 1144 | AAI07272.1 | |
| BC107272.1 | 5 | 1145 | AAI07273.1 |

Genomic Map