Assay Details
Target Gene Details
Entrez Gene ID: | 319713 |
Gene Name: | actin binding LIM protein family, member 3 |
Gene Aliases: |
AW990719, D930036B08Rik |
Location: |
Chr.18:61799392-61911857 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ablim3 | NM_001164491.1 | 4 | 508 | NP_001157963.1 |
| NM_198649.3 | 3 | 827 | NP_941051.2 | |
| AK086538.1 | 3 | 607 | ||
| AK173040.1 | 3 | 385 | BAD32318.1 | |
| BC060275.1 | 4 | 479 | AAH60275.1 | |
| BC094229.1 | 3 | 801 | AAH94229.1 |
More Information
Set Membership: |
|

Genomic Map