Assay Details
Target Gene Details
Entrez Gene ID: | 319713 |
Gene Name: | actin binding LIM protein family, member 3 |
Gene Aliases: |
AW990719, D930036B08Rik |
Location: |
Chr.18:61799392-61911857 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ablim3 | NM_001164491.1 | 5 | 668 | NP_001157963.1 |
| NM_198649.3 | 4 | 987 | NP_941051.2 | |
| AK086538.1 | 4 | 767 | ||
| AK173040.1 | 4 | 545 | BAD32318.1 | |
| BC060275.1 | 5 | 639 | AAH60275.1 | |
| BC094229.1 | 4 | 961 | AAH94229.1 |
More Information
Set Membership: |
|

Genomic Map