Assay Details
Target Gene Details
Entrez Gene ID: | 240119 |
Gene Name: | beta galactoside alpha 2,6 sialyltransferase 2 |
Gene Aliases: |
C230064G14Rik, St6galII, mKIAA1877 |
Location: |
Chr.17:55444358-55514581 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6gal2 | NM_172829.2 | NP_766417.1 | ||
| AK054230.1 | 1 | 346 | ||
| AK082566.1 | BAC38534.1 |
More Information
Set Membership: |
|

Genomic Map