Assay Details
Target Gene Details
Entrez Gene ID: | 106757 |
Gene Name: | catsper channel auxiliary subunit delta |
Gene Aliases: |
4921529N20Rik, 4933402B14Rik, 619718, AW045609, Gm6095, Tmem146 |
Location: |
Chr.17:56626896-56664460 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Catsperd | NM_175350.3 | 20 | 1893 | NP_780559.2 |
| AK077082.1 | 11 | 954 | BAC36602.1 | |
| BC115864.1 | 9 | 846 | ||
| BC116633.1 | 10 | 960 | ||
| HQ441159.1 | 20 | 1885 | ||
| HQ441160.1 | 17 | 1781 |

Genomic Map