Assay Details
Target Gene Details
Entrez Gene ID: | 224826 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 2 |
Gene Aliases: |
9930021A08Rik, AI462103, AW540746, E130209G04Rik, mKIAA0349 |
Location: |
Chr.17:46928291-47010775 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubr2 | NM_001177374.1 | 30 | 3697 | NP_001170845.1 |
| NM_146078.3 | 30 | 3697 | NP_666190.2 | |
| AK122254.2 | 28 | 4526 | BAC65536.3 | |
| AK143202.1 | 30 | 3697 | BAE25301.1 | |
| AK147445.1 | 30 | 3686 | BAE27917.1 | |
| AY061885.1 | 30 | 3384 | AAL32102.1 | |
| AY280958.1 | 30 | 3412 | AAQ17202.1 | |
| BC026391.1 | 4 | 450 | AAH26391.1 | |
| BC031403.1 | 15 | 1491 | AAH31403.1 | |
| BC075642.1 | 31 | 3511 | AAH75642.1 |
More Information
Set Membership: |
|

Genomic Map