Assay Details
Target Gene Details
Entrez Gene ID: | 69456 |
Gene Name: | COMM domain containing 10 |
Gene Aliases: |
2310003A05Rik, AU019438, DRWMS2 |
Location: |
Chr.18:46958785-47087995 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Commd10 | NM_178377.4 | NP_848464.1 | ||
| AB052770.1 | BAC53803.1 | |||
| AK159099.1 | BAE34815.1 | |||
| AK166907.1 | BAE39108.1 | |||
| BC034879.1 | AAH34879.1 |

Genomic Map