Assay Details
Target Gene Details
Entrez Gene ID: | 240119 |
Gene Name: | beta galactoside alpha 2,6 sialyltransferase 2 |
Gene Aliases: |
C230064G14Rik, St6galII, mKIAA1877 |
Location: |
Chr.17:55444358-55514581 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6gal2 | NM_172829.2 | 4 | 1236 | NP_766417.1 |
| AK082566.1 | 4 | 1237 | BAC38534.1 | |
| AK129462.1 | 2 | 168 | BAC98272.1 | |
| BC125586.1 | 3 | 1106 | ||
| BC138142.1 | 3 | 1106 |
More Information
Set Membership: |
|

Genomic Map