Assay Details
Target Gene Details
Entrez Gene ID: | 106757 |
Gene Name: | catsper channel auxiliary subunit delta |
Gene Aliases: |
4921529N20Rik, 4933402B14Rik, 619718, AW045609, Gm6095, Tmem146 |
Location: |
Chr.17:56626896-56664460 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Catsperd | NM_175350.3 | NP_780559.2 | ||
| AK077082.1 | BAC36602.1 | |||
| BC115864.1 | ||||
| BC116633.1 | ||||
| HQ441159.1 | ||||
| HQ441160.1 |

Genomic Map