Assay Details
Target Gene Details
Entrez Gene ID: | 83672 |
Gene Name: | synaptotagmin-like 3 |
Gene Aliases: |
A430092N21, Slp3, Slp3-a, Slp3-b, mFLJ00307 |
Location: |
Chr.17:6658233-6738070 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sytl3 | NM_031395.2 | NP_113572.1 | ||
| NM_183368.2 | NP_899224.2 | |||
| NM_183370.2 | NP_899226.2 | |||
| AB057758.1 | BAB41086.1 | |||
| AB057764.1 | BAB41092.1 | |||
| AB057765.1 | BAB41093.1 | |||
| AK131174.1 | BAD21424.1 | |||
| BC022608.1 | AAH22608.1 | |||
| BC106102.1 | AAI06103.1 | |||
| BC118936.2 | ||||
| BC152315.1 |

Genomic Map