Assay Details
Target Gene Details
Entrez Gene ID: | 70950 |
Gene Name: | coiled coil domain containing 178 |
Gene Aliases: |
4921528I01Rik |
Location: |
Chr.18:21810891-22171442 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc178 | NM_027616.3 | 19 | 2134 | NP_081892.2 |
| AK029534.1 | 19 | 2135 | BAC26501.1 | |
| BC140993.1 | 19 | 2172 | ||
| BC145066.1 | 19 | 2172 | ||
| BC145069.1 | 19 | 2179 |

Genomic Map