Assay Details
Target Gene Details
Entrez Gene ID: | 268980 |
Gene Name: | striatin, calmodulin binding protein |
Gene Aliases: |
AU022939, D130055P19 |
Location: |
Chr.17:78649908-78737191 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strn | NM_011500.2 | NP_035630.2 | ||
| AJ223777.1 | CAA11545.1 | |||
| AK051549.1 | BAC34670.1 | |||
| BC090968.1 | AAH90968.1 | |||
| BC150727.1 |

Genomic Map