Assay Details
Target Gene Details
Entrez Gene ID: | 78058 |
Gene Name: | RIKEN cDNA 4930578E11 gene |
Gene Aliases: |
- |
Location: |
Chr.18:29396716-29496416 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930578E11Rik | NR_045391.1 | 7 | 1515 | |
| AK132929.1 | 7 | 1516 |

Genomic Map