Assay Details
Target Gene Details
Entrez Gene ID: | 106877 |
Gene Name: | actin filament associated protein 1-like 1 |
Gene Aliases: |
AI173486 |
Location: |
Chr.18:61730262-61786662 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Afap1l1 | NM_178928.4 | 8 | 908 | NP_849259.2 |
| AK035227.1 | 8 | 908 | BAC28988.1 | |
| AK053714.1 | 8 | 907 | BAC35486.1 | |
| BC138923.1 | 8 | 869 |

Genomic Map