Assay Details
Target Gene Details
Entrez Gene ID: | 56424 |
Gene Name: | STIP1 homology and U-Box containing protein 1 |
Gene Aliases: |
0610033N24Rik, 2210017D18Rik, 2310040B03Rik, AW046544, Chip |
Location: |
Chr.17:25830636-25833361 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stub1 | NM_019719.3 | NP_062693.1 | ||
| AF129086.1 | AAD33401.1 | |||
| AK002752.1 | BAB22329.1 | |||
| AK004464.1 | BAB23315.1 | |||
| AK045776.1 | BAC32489.1 | |||
| AK158595.1 | BAE34573.1 | |||
| AK166630.1 | BAE38905.1 | |||
| AK183097.1 | ||||
| AK209265.1 | ||||
| BC027427.1 | AAH27427.1 | |||
| BC038939.1 | AAH38939.1 |
More Information
Set Membership: |
|

Genomic Map