Assay Details
Target Gene Details
Entrez Gene ID: | 72106 |
Gene Name: | jumonji domain containing 8 |
Gene Aliases: |
2610003J06Rik |
Location: |
Chr.17:25829043-25831843 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jmjd8 | NM_028101.4 | 9 | 1490 | NP_082377.2 |
| AK011308.1 | 9 | 1487 | BAB27534.1 | |
| AK148079.1 | 9 | 1468 | BAE28331.1 | |
| AK155810.1 | 9 | 1492 | BAE33443.1 | |
| AK172073.1 | 9 | 1487 | BAE42811.1 | |
| BC010800.1 | 9 | 1452 | AAH10800.1 |
Target Gene Details
Entrez Gene ID: | 56424 |
Gene Name: | STIP1 homology and U-Box containing protein 1 |
Gene Aliases: |
0610033N24Rik, 2210017D18Rik, 2310040B03Rik, AW046544, Chip |
Location: |
Chr.17:25830636-25833361 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stub1 | NM_019719.3 | NP_062693.1 | ||
| AF129086.1 | AAD33401.1 | |||
| AK002752.1 | BAB22329.1 | |||
| AK004464.1 | BAB23315.1 | |||
| AK045776.1 | BAC32489.1 | |||
| AK158595.1 | BAE34573.1 | |||
| AK166630.1 | BAE38905.1 | |||
| AK177504.1 | ||||
| AK184249.1 | ||||
| AK197887.1 | ||||
| AK213284.1 | 1 | 304 | ||
| BC027427.1 | AAH27427.1 | |||
| BC038939.1 | AAH38939.1 |
More Information
Set Membership: |
|

Genomic Map