Assay Details
Target Gene Details
Entrez Gene ID: | 21678 |
Gene Name: | TEA domain family member 3 |
Gene Aliases: |
DTEF-1, ETFR-1, TEAD-3, TEF-5, Tcf13r2 |
Location: |
Chr.17:28331671-28350805 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tead3 | NM_001098226.3 | NP_001091696.2 | ||
| NM_001204156.1 | NP_001191085.1 | |||
| NM_011566.4 | NP_035696.3 | |||
| AF002670.1 | AAC12263.1 | |||
| AK087649.1 | ||||
| AK143551.1 | ||||
| BC021364.1 | AAH21364.1 | |||
| BC027383.1 | AAH27383.1 | |||
| BC039984.1 | AAH39984.1 | |||
| CB182196.1 | ||||
| D87963.1 | BAA13516.1 | |||
| Y10027.1 | CAA71136.1 | |||
| Y16611.1 | CAA76315.2 |
More Information
Set Membership: |
|

Genomic Map