Assay Details
Target Gene Details
Entrez Gene ID: | 225187 |
Gene Name: | ankyrin repeat domain 29 |
Gene Aliases: |
G630054C21, G630054C21Rik, Gm95 |
Location: |
Chr.18:12252357-12305720 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd29 | NM_001190371.1 | 9 | 866 | NP_001177300.1 |
| AK090336.1 | 9 | 819 | ||
| AK162301.1 | 4 | 294 | BAE36839.1 |

Genomic Map