Assay Details
Target Gene Details
Entrez Gene ID: | 21785 |
Gene Name: | trefoil factor 2 (spasmolytic protein 1) |
Gene Aliases: |
SP, mSP |
Location: |
Chr.17:31141062-31144282 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tff2 | NM_009363.3 | NP_033389.2 | ||
| AK003093.1 | BAB22560.1 | |||
| BC050086.1 | AAH50086.1 | |||
| BC087889.1 | AAH87889.1 | |||
| X51697.1 | CAA35994.1 |
More Information
Set Membership: |
|

Genomic Map