Assay Details
Target Gene Details
Entrez Gene ID: | 21785 |
Gene Name: | trefoil factor 2 (spasmolytic protein 1) |
Gene Aliases: |
SP, mSP |
Location: |
Chr.17:31141062-31144282 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tff2 | NM_009363.3 | 2 | 231 | NP_033389.2 |
| AK003093.1 | 2 | 232 | BAB22560.1 | |
| BC050086.1 | 2 | 192 | AAH50086.1 | |
| BC087889.1 | 2 | 209 | AAH87889.1 | |
| X51697.1 | 2 | 196 | CAA35994.1 |
More Information
Set Membership: |
|

Genomic Map