Assay Details
Target Gene Details
Entrez Gene ID: | 240479 |
Gene Name: | family with sequence similarity 69, member C |
Gene Aliases: |
B230399E16Rik |
Location: |
Chr.18:84718987-84741731 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam69c | NM_173770.4 | NP_776131.1 | ||
| AK046504.1 | BAC32758.1 |

Genomic Map