Assay Details
Target Gene Details
Entrez Gene ID: | 27279 |
Gene Name: | tumor necrosis factor receptor superfamily, member 12a |
Gene Aliases: |
AI255180, C87282, Fn14, HPIP, TWEAK-R, TweakR |
Location: |
Chr.17:23675445-23677449 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf12a | NM_001161746.1 | NP_001155218.1 | ||
| NM_013749.2 | NP_038777.2 | |||
| AF156164.1 | AAF07882.1 | |||
| AK005382.1 | BAB23989.1 | |||
| AK005530.1 | BAB24101.1 | |||
| AK160136.1 | BAE35650.1 | |||
| BC025860.1 | AAH25860.1 | |||
| DV655788.1 |
More Information
Set Membership: |
|

Genomic Map