Assay Details
Target Gene Details
Entrez Gene ID: | 381085 |
Gene Name: | TBC1 domain family, member 22B |
Gene Aliases: |
BC045600 |
Location: |
Chr.17:29549788-29606894 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d22b | NM_198647.1 | NP_941049.1 | ||
| BC045600.1 | AAH45600.1 |
More Information
Set Membership: |
|

Genomic Map