Assay Details
Target Gene Details
Entrez Gene ID: | 17229 |
Gene Name: | tryptase beta 2 |
Gene Aliases: |
AV011504, MMCP-6, Mcp-6, Mcp6, Mcpt6 |
Location: |
Chr.17:25366303-25368096 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpsb2 | M57626.1 | AAA39988.1 |

Genomic Map