Assay Details
Target Gene Details
Entrez Gene ID: | 70425 |
Gene Name: | casein kinase 1, gamma 3 |
Gene Aliases: |
3300002K07Rik, C330049O21Rik |
Location: |
Chr.18:53862082-53955684 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1g3 | NM_152809.2 | NP_690022.2 | ||
| AK032675.1 | BAC27983.1 | |||
| AK218781.1 | ||||
| BC033601.1 |
More Information
Set Membership: |
|

Genomic Map