Assay Details
Target Gene Details
Entrez Gene ID: | 50850 |
Gene Name: | spastin |
Gene Aliases: |
Spg4, mKIAA1083 |
Location: |
Chr.17:74337454-74391115 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spast | NM_001162870.1 | NP_001156342.1 | ||
| NM_016962.2 | NP_058658.2 | |||
| AJ246002.1 | CAB60143.1 | |||
| AK007793.1 | BAB25259.1 | |||
| AK129282.1 | BAC98092.1 | |||
| BC046286.1 | AAH46286.1 |
More Information
Set Membership: |
|

Genomic Map