Assay Details
Target Gene Details
Entrez Gene ID: | 240057 |
Gene Name: | synaptic Ras GTPase activating protein 1 homolog (rat) |
Gene Aliases: |
Gm1963, Syngap |
Location: |
Chr.17:26941408-26972434 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syngap1 | NM_001281491.1 | NP_001268420.1 |
More Information
Set Membership: |
|

Genomic Map