Assay Details
Target Gene Details
Entrez Gene ID: | 381085 |
Gene Name: | TBC1 domain family, member 22B |
Gene Aliases: |
BC045600 |
Location: |
Chr.17:29549788-29606894 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d22b | NM_198647.1 | 9 | 1174 | NP_941049.1 |
| BC045600.1 | 9 | 1174 | AAH45600.1 | |
| BC060066.1 | 4 | 446 | AAH60066.1 |
Target Gene Details
Entrez Gene ID: | 74092 |
Gene Name: | TBC1 domain family, member 22B, opposite strand |
Gene Aliases: |
0610038L08Rik |
Location: |
Chr.17:29578816-29596305 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d22bos | NR_045447.1 | |||
| AK002805.1 |
More Information
Set Membership: |
|

Genomic Map