Assay Details
Target Gene Details
Entrez Gene ID: | 17229 |
Gene Name: | tryptase beta 2 |
Gene Aliases: |
AV011504, MMCP-6, Mcp-6, Mcp6, Mcpt6 |
Location: |
Chr.17:25366303-25368096 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpsb2 | NM_010781.3 | 4 | 551 | NP_034911.3 |
| BC024374.1 | 4 | 551 | AAH24374.1 | |
| L31853.1 | 4 | 545 | AAA39725.1 | |
| M57626.1 | 5 | 573 | AAA39988.1 |
More Information
Set Membership: |
|

Genomic Map