Assay Details
Target Gene Details
Entrez Gene ID: | 67664 |
Gene Name: | ring finger protein 125 |
Gene Aliases: |
4930553F04Rik |
Location: |
Chr.18:20944625-20987355 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf125 | NM_026301.2 | NP_080577.1 | ||
| AB259692.1 | ||||
| AK006569.1 | BAB24656.1 | |||
| AK016103.1 | ||||
| AK050395.1 | BAC34234.1 | |||
| BC094290.1 | AAH94290.1 | |||
| BC116883.1 | ||||
| BC116887.1 |

Genomic Map